Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM10 Antibody (MM0077-6D31), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212014AF405
Description
ADAM10 Monoclonal specifically detects ADAM10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ADAM10 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADAM10 | |
Human recombinant ADAM10 (extracellular domain) | |
0.1 mL | |
Primary | |
In Western blots, this antibody shows weak cross-reactivity with recombinant mouse ADAM10 and human ADAM8, 9, 15, and 17. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0077-6D31 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
Mouse | |
Protein G purified | |
Apoptosis, Signal Transduction | |
102 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction