Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM10 Rat anti-Mouse, Alexa Fluor 647, Clone: 139712, R&D Systems™

Rat MonoclonalAntibody

Manufacturer:  R&D Systems FAB946R100UG

Catalog No. FAB946R100


Add to cart
This item is not returnable. View return policy

Description

Description

ADAM10 Monoclonal antibody specifically detects ADAM10 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

ADAM10
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
O35598
ADAM10
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM10 Thr215-Glu673 Accession # O35598
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse ADAM10 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) rhADAM10 is observed and no cross-reactivity with rhADAM8, recombinant mouse (rm) ADAM9, rmADAM15, rhBACE, or rhTACE is observed.
Flow Cytometry
139712
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian, Kuzbanian protein homolog, MADM, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease
Rat
IgG2a
RUO
Primary
102
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)