Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548657

Catalog No. PA548657

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probable ligand for integrin in the brain. This is a non catalytic metalloprotease-like protein.
Specifications

Specifications

ADAM11
Polyclonal
PBS with 0.09% sodium azide
O75078
Disintegrin and metalloproteinase domain-containing protein 11, ADAM 11, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein, MDC, ADAM11, MDC
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4185
Western Blot
Unconjugated
ADAM11
Liquid
ADAM11
KLH conjugated synthetic peptide between 200-228 amino acids from the N-terminal region of human ADAM11.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit