Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM12 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA5121158

Encompass

Catalog No. PIPA5121158


Add to Cart
This item is not returnable. View return policy

Description

Description

Positive test controls include: Mouse brain, Mouse thymus, Mouse fat, Mouse ovary, Rat brain. The target is usually found in the following locations: Cell membrane, Secreted, Single-pass type I membrane protein.

ADAM12 Polyclonal antibody specifically detects ADAM12 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

ADAM12
Polyclonal
Unconjugated
ADAM12
a disintegrin and metallopeptidase domain 12 (meltrin alpha); a disintegrin and metalloprotease domain 12; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 12 (meltrin alpha); ADAM; ADAM 12; ADAM metallopeptidase domain 12; Adam12; ADAM12-OT1; ADAMs; CAR10; Disintegrin and metalloproteinase domain-containing protein 12; MCMP; MCMPMltna; meltrin alpha; meltrin-alpha; metalloendopeptidases; metalloprotease-disintegrin 12 transmembrane; mKIAA4001; MLTN; Mltna; PRO545; RP11-295J3.5; UNQ346; UNQ346/PRO545
Recombinant fusion protein containing a sequence corresponding to amino acids 589-738 of human ADAM12 (NP_067673.2).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
2.69 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43184, Q61824
Rabbit
Antigen affinity chromatography
RUO
11489, 679837, 8038
-20° C, Avoid Freeze/Thaw Cycles
Liquid
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit