Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM12 Rabbit anti-Human, Clone: 737, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | ADAM12 |
---|---|
Clone | 737 |
Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
ADAM12 Monoclonal specifically detects ADAM12 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ADAM12 | |
Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
8038 | |
IgG | |
Protein A purified |
737 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
a disintegrin and metalloproteinase domain 12 (meltrin alpha), ADAM 12, ADAM metallopeptidase domain 12, disintegrin and metalloproteinase domain-containing protein 12, EC 3.4.24, EC 3.4.24.58, MCMP, MCMPMltna, Meltrin-alpha, MLTNA, MLTNEC 3.4.24.- | |
Recombinant Human ADAM12 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title