Learn More
Novus Biologicals™ ADAM15 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2794825PLATES
 View more versions of this product
Catalog No. NB010004
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ADAM15.
Specifications
Specifications
HRP
Antibody Pairs
47 pg/mL
8751
RUO
ADAM15
5 Plates
46.88-3000 pg/mL
ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ADAM15.
Sandwich ELISA
  • Standard: Each vial contains 96 ng of recombinant ADAM15.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-ADAM15 monoclonal antibody(in PBS, pH 7.4).
  • Detection Antibody: 0.5 mg/mL rabbit anti-ADAM15 monoclonal antibodyconjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Storage is content dependent.
    Rabbit
    Human
    ADAM15
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3 ng/mL is recommended.