Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM15 Rabbit anti-Human, PerCP, Clone: 7, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289546PCP

Catalog No. NB067251


Add to cart

Description

Description

ADAM15 Monoclonal antibody specifically detects ADAM15 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

ADAM15
7
Flow Cytometry
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
PerCP
PBS
ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin
Recombinant Human ADAM15 protein
Protein A purified
RUO
Primary
8751
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit