Learn More
ADAM17 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 202591AP
Catalog No. 50-173-2800
Add to cart
Description
Description
This antibody is specific to ADAM17.
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biologic processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene functions as a tumor necrosis factor-alpha converting enzyme; binds mitotic arrest deficient 2 protein; and also plays a prominent role in the activation of the Notch signaling pathway.
Specifications
Specifications
ADAM17
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P78536
ADAM 17, ADAM17, CD156b, cSVP, Snake venom like protease, TACE, TNF alpha convertase, TNF alpha converting enzyme
Rabbit
IgG
150 μL
-20°C
Primary
6868
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ADAM17
Liquid
ADAM17
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human