Learn More
ADAM19 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 222161AP
Catalog No. 50-173-3776
Add to cart
Description
Description
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a type I transmembrane protein and serves as a marker for dendritic cell differentiation. It has also been demonstrated to be an active metalloproteinase, which may be involved in normal physiological and pathological processes such as cells migration, cell adhesion, cell-cell and cell-matrix interactions, and signal transduction.
Specifications
Specifications
ADAM19
0.78 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35674, Q9H013
ADAM 19, ADAM19, FKSG34, MADDAM, Meltrin beta, MLTNB
Rabbit
IgG
150 μL
-20°C
Primary
11492, 8728
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ADAM19
Liquid
ADAM19
ADAM19 Fusion Protein Ag17544
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse