Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM22 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen ADAM22
Immunogen Synthetic peptides corresponding to Adam22 (a disintegrin and metallopeptidase domain 22) The peptide sequence was selected from the N terminal of mouse Adam22 (NP_001007222). Peptide sequence AISENPLITLREFMKYRRDFIKEKADAVHLFSGSQFESSRSGAAYIGGIC.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16901520 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16901520UL
20ul Each for $149.00
Add to cart
 
NBP169015 View Documents Novus BiologicalsSupplier Diversity Partner
NBP169015
100 ul Each for $329.00
Add to cart
 
Description

Description

ADAM22 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

ADAM22
Rabbit
Polyclonal
Unconjugated
53616
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to Adam22 (a disintegrin and metallopeptidase domain 22) The peptide sequence was selected from the N terminal of mouse Adam22 (NP_001007222). Peptide sequence AISENPLITLREFMKYRRDFIKEKADAVHLFSGSQFESSRSGAAYIGGIC.
Primary
IgG
ADAM 22, ADAM metallopeptidase domain 22, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit