Learn More
ADAM28, Mouse anti-Human, Alexa Fluor 594, Clone: 1004266, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB93252T100UG
Catalog No. FAB93252T10
Add to cart
This item is not returnable. View return policy
Description
Description
ADAM28 Monoclonal antibody specifically detects ADAM28 in Human Samples. It is validated for Flow Cytometry.
Specifications
Specifications
ADAM28
1004266
Flow Cytometry 0.25-1 ug/10^6 cells
Q0UKQ2
a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, Dtgn1, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCL, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L, Protein-L
Chinese Hamster Ovary cell line, CHO-derived human ADAM28, Leu72 & Leu191-Ala623,, Accession # Q0UKQ2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10863
Human
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG1
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human ADAM28 in direct ELISAs.