Learn More
ADAM29 Mouse anti-Human, Clone: 3A6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524562
Catalog No. PIMA524562
Add to cart
Description
Description
Peptide Sequence: GMNHDEDTCR CSQPRCIMHE GNPPITKFSN CSYGDFWEYT VERTKCLLET VHTKDIFNVK
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene is highly expressed in testis and may be involved in human spermatogenesis. Alternative splicing results in multiple transcript variants that encode the same protein.
Specifications
Specifications
ADAM29
Monoclonal
Unconjugated
ADAM29
Liquid
ADAM29
ADAM29 (NP_055084, 339 a.a. ∽ 398 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
3A6
PBS with no preservative; pH 7.4
Q9UKF5
CT73, svph1
Mouse
IgG1, kappa
100 μg
-20° C
Primary
11086