Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 221471AP150UL
Description
This antibody is specific to ADAM5.
ADAM5 (putative disintegrin and metalloproteinase domain-containing protein 5) is also named as ADAM5P, TMDC2(tMDCII). The tMDC II, abundantly expressed, sperm surface MDC proteins identified in rodent and macaque systems, is also nonfunctional in the human. It has 7 isoforms produced by alternative splicing. It is expressed in testis highly by northern blot.Specifications
ADAM5 | |
Polyclonal | |
Unconjugated | |
ADAM5 | |
ADAM5, ADAM5P, TMDCII | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
255926 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6NVV9 | |
ADAM5 | |
ADAM5 Fusion Protein Ag17590 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction