Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM7 Mouse anti-Human, Mouse, Rat, Clone: 4D1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275396

Catalog No. NBP275396


Add to cart

Description

Description

ADAM7 Monoclonal antibody specifically detects ADAM7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

ADAM7
4D1
Unconjugated
TBS (pH7.4), 0.5%BSA, 50%Glycerol with 0.05% Sodium Azide
a disintegrin and metalloproteinase domain 7, ADAM 7, ADAM metallopeptidase domain 7, disintegrin and metalloproteinase domain-containing protein 7, EAPI, epididymal apical protein I, GP83, GP-83, Sperm maturation-related glycoprotein GP-83
Mouse
IgG1
Protein G purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin
Purified
ADAM7
Recombinant protein
85 kDa
100 μL
Primary
8756.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit