Learn More
ADAM8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA587946
Description
Immunogen sequence: IAPSRPWALM EQYEVVLPWR LPGPRVRRAL PSHLGLHPER VSYVLGATGH NFTLHLRKNR DLLGSGYTET YTAANGSEVT EQPRGQDHCF YQGHVEGYPD SAASLSTCAG LRGFFQVGSD LHLIEPLDEG GEGGRHAVYQ AEHLLQTAGT CGVSDDSLGS LLGPRTAAVF RPRPGDSLPS RETRYVELYV VVDNAEFQML GSEAAVRHRV LEVVNHVDKL YQKLNFRVVL VGLEIWNSQD RFHVSPDPSV TLENLLTWQA RQRTRRHLHD NVQLITGVDF TGTT; Positive Samples: BxPC-3, NIH/3T3; Cellular Location: Membrane, Single-pass type I membrane protein
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene may be involved in cell adhesion during neurodegeneration, and it is thought to be a target for allergic respiratory diseases, including asthma. Alternative splicing results in multiple transcript variants.Specifications
ADAM8 | |
Polyclonal | |
Unconjugated | |
ADAM8 | |
ADAM8 | |
Recombit fusion protein containing a sequence corresponding to amino acids 17-300 of human ADAM8 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
5.68 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78325, Q05910 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
101, 11501 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.