Learn More
ADAM9 Rat anti-Mouse, DyLight 350, Clone: RM0071-7D10, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP121692UV
Catalog No. NBP121692UV
Add to cart
Description
Description
ADAM9 Monoclonal antibody specifically detects ADAM9 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
ADAM9
RM0071-7D10
Western Blot
Purified
ADAM9
Mouse recombinant ADAM9 EC-domain
Protein G purified
Signal Transduction
Primary
8754
Mouse
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMPMDC9KIAA0021Mltng, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant ADAM9 Extracellular domain.