Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM9 Rat anti-Mouse, DyLight 755, Clone: RM0071-7D10, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP121692IR
Description
ADAM9 Monoclonal antibody specifically detects ADAM9 in Mouse samples. It is validated for Western Blot.Specifications
ADAM9 | |
RM0071-7D10 | |
Western Blot | |
Purified | |
ADAM9 | |
Mouse recombinant ADAM9 EC-domain | |
Protein G purified | |
Signal Transduction | |
Primary | |
8754 | |
Mouse |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMPMDC9KIAA0021Mltng, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase | |
Rat | |
IgG2 | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant ADAM9 Extracellular domain. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title