Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS10 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6588R

Description
ADAMTS (a disintegrin and metalloproteinase domain with Thrombospondin type-1 modules) is a family of zinc-dependent proteases that are implicated in a variety of normal and pathological conditions, including arthritis and cancer (1-3). Embryogenesis, morphological growth changes, inflammation, tumor invasion and metastasis involve the breakdown and remodeling of the extracellular matrix. This degradation is due to the family of enzymes known as the matrix metalloproteinases (MMPs), which are related to ADAMTS subtypes. ADAMTS protein family members contain an amino-terminal propeptide domain, a metalloproteinase domain, a disintegrin-like domain and a carboxy-terminus that contains a varying number of thrombospondin type-1 (TSP-1) motifs (1-3). ADAMTS genes are primarily expressed in fetal tissues, including the lung, kidney and liver (1-3). The human ADAMTS10 gene maps to chromosome 19p13.3 (4). The human ADAMTS19 gene maps to chromosome 5q31 (1,5).Specifications
ADAMTS10 | |
Polyclonal | |
Unconjugated | |
ADAMTS10 | |
9430006O18; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 10; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 10; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 10; a disintegrin-like and metalloprotease with thrombospondin type 1 motif, 10; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 10; ADAM metallopeptidase with thrombospondin type 1 motif, 10; ADAMs; ADAM-TS 10; ADAMTS10; Adam-ts10; Adamts-10; AW045948; metalloendopeptidases; WMS; WMS1; zinc metalloendopeptidase; Znmp | |
Rabbit | |
Protein A | |
RUO | |
81794 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9H324 | |
ADAMTS10 | |
KLH conjugated synthetic peptide derived from human ADAMTS10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction