Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTS14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570703
Description
This target displays homology in the following species: Cow: 79%; Dog: 93%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 86%; Rat: 100%; Zebrafish: 82%.
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. This gene is highly similar to two family members, ADAMTS2 and ADAMTS3, in its sequence and gene structure, and the encoded protein shares the aminoprocollagen peptidase activity with the protein products encoded by ADAMTS2 and ADAMTS3. Various transcript variants of this gene have been identified. They result from the use of two different promoters and transcription initiation sites as well as alternative splicing sites. The full length nature of some transcripts has not been defined.
Specifications
ADAMTS14 | |
Polyclonal | |
Unconjugated | |
ADAMTS14 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 14; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 14; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 14; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 14; ADAM metallopeptidase with thrombospondin type 1 motif, 14; ADAMs; ADAM-TS 14; ADAMTS14; ADAM-TS14; Adamts-14; ATS14; FLJ32820; metalloendopeptidases; TS14 | |
Rabbit | |
Affinity chromatography | |
RUO | |
140766 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WXS8 | |
ADAMTS14 | |
synthetic peptide directed towards the N-terminal region of human ADAMTS14. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction