Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550599
Description
The antibody detects endogenous levels of total ADAMTS17 protein.
ADAMTS17 Polyclonal antibody specifically detects ADAMTS17 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ADAMTS17 | |
Polyclonal | |
Unconjugated | |
ADAMTS17 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 17; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 17; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 17; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 17; ADAM metallopeptidase with thrombospondin type 1 motif, 17; ADAMs; ADAM-TS 17; ADAMTS17; ADAM-TS17; ADAMTS-17; ATS17; AU023434; disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 17; FLJ16363; FLJ32769; metalloendopeptidases | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170691, 233332 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8TE56 | |
ADAMTS17 | |
Synthetic peptide corresponding to a region derived from internal residues of human ADAM metallopeptidase with thrombospondin type 1 motif, 17 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction