Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550539

Catalog No. PA550539


Add to cart

Description

Description

The antibody detects endogenous levels of total ADAMTS2 protein.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene excises the N-propeptide of type I, type II and type V procollagens. Mutations in this gene cause Ehlers-Danlos syndrome type VIIC, a recessively inherited connective-tissue disorder. Alternative splicing results in two transcript variants. The short transcript encodes a protein which has no significant procollagen N-peptidase activity.
Specifications

Specifications

ADAMTS2
Polyclonal
Unconjugated
ADAMTS2
Liquid
ADAMTS2
Synthetic peptide corresponding to residues near the C terminal of human ADAM metallopeptidase with thrombospondin type 1 motif, 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
O95450
ADAM-TS2, ADAMTS-3, NPI, PCINP, PCPNI, hPCPNI
Rabbit
IgG
100 μL
-20°C
Primary
9509
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit