Learn More
ADAMTS4 Mouse anti-Human, Clone: OTI6F8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805208
Catalog No. 50-166-7322
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the ADAMTS protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene lacks a C-terminal TS motif. It is responsible for the degradation of aggrecan, a major proteoglycan of cartilage, and brevican, a brain-specific extracellular matrix protein. The cleavage of aggrecan and brevican suggests key roles of this enzyme in arthritic disease and in the central nervous system, potentially, in the progression of glioma.
Specifications
Specifications
ADAMTS4
Monoclonal
Unconjugated
ADAMTS4
Lyophilized
ADAMTS4
Human recombit protein fragment corresponding to amino acids 296-577 of human ADAMTS4 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI6F8
PBS with 8% trehalose and no preservative; pH 7.3
O75173
ADAMTS-2, ADAMTS-4, ADMP-1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9507