Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550540
Description
The antibody detects endogenous levels of total ADAMTS6 protein.
ADAMTS6 Polyclonal antibody specifically detects ADAMTS6 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
ADAMTS6 | |
Polyclonal | |
Unconjugated | |
ADAMTS6 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 6; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 6; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 6; ADAM metallopeptidase with thrombospondin type 1 motif, 6; ADAMs; ADAM-TS 6; ADAMTS6; ADAM-TS6; ADAMTS-6; metalloendopeptidases | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11174 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UKP5 | |
ADAMTS6 | |
Synthetic peptide corresponding to a region derived from internal residues of human ADAM metallopeptidase with thrombospondin type 1 motif, 6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction