Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS8 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2H5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271677JF549

Catalog No. N271677J549


Add to cart

Description

Description

ADAMTS8 Monoclonal antibody specifically detects ADAMTS8 in Human samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

ADAMTS8
OTI2H5
Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
ADAMTS8
Full length recombinant protein of human ADAMTS8 (NP_008968) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication
Primary
11095.0
Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 8, ADAM metallopeptidase with thrombospondin type 1 motif, 8, ADAMTS-8, ADAM-TS8A disintegrin and metalloproteinase with thrombospondin motifs 8, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82, FLJ41712, METH-2, METH2ADAM-TS 8, METH-8
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit