Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTSL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550306
Description
The antibody detects endogenous levels of total ADAMTSL1 protein.
Punctin (ADAMTSL-1) is a secreted molecule, expressed in adult skeletal muscle, resembling members of the ADAMTS family of proteases, containing four thrombospondin type I repeats. Human punctin cleaves aggrecan and is thereby classified as an aggrecanase. Like the ADAMTS proteases, each domain in punctin has an even number of cysteine residues suggesting that each domain may have internal disulfide bonds and that punctin consists of a series of independently folded and disulfide-bonded domains.
Specifications
ADAMTSL1 | |
Polyclonal | |
Unconjugated | |
ADAMTSL1 | |
5930437A14Rik; 6720426B09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 1; ADAM-TS related protein 1; Adamtsl1; ADAMTSL-1; ADAMTS-like 1; ADAMTS-like protein 1; ADAMTSR1; C9orf94; DKFZp686L03130; FLJ35283; FLJ41032; FLJ46891; metalloendopeptidases; MGC118803; MGC118805; MGC40193; PUNCTIN; Punctin-1; RP11-220B22.2; UNQ528/PRO1071 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92949 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N6G6 | |
ADAMTSL1 | |
Fusion protein corresponding to a region derived from internal residues of human ADAMTS-like 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction