Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTSL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552269
Description
Immunogen sequence: ISSPPPILEN PTPEPPVPQL QPEILRVEPP LAPAPRPART PGTLQRQVRI PQMPAPPHPR TPLGSPAAYW KRVGHSACSA SCGKGVWRPI FLCISRESGE ELDERS Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.
This gene is a member of ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs)-like gene family and encodes a protein with protein with seven thrombospondin type 1 repeats. The thrombospondin type 1 repeat domain is found in many proteins with diverse biological functions including cellular adhesion, angiogenesis, and patterning of the developing nervous system. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
ADAMTSL4 | |
Polyclonal | |
Unconjugated | |
ADAMTSL4 | |
a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 4; adamtsl4; ADAMTSL-4; adamtsl4 {ECO:0000312; ADAMTS-like 4; ADAMTS-like protein 4; ECTOL2; EMBL:AAH99119.1}; metalloendopeptidases; PP1396; thrombospondin repeat containing 1; thrombospondin repeat protein 1; thrombospondin repeat-containing protein 1; Tsrc1; UNQ2803/PRO34012 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54507 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UY14 | |
ADAMTSL4 | |
Recombinant protein corresponding to Human ADAMTSL4. Recombinant protein control fragment (Product #RP-89166). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction