Learn More
Tocris Bioscience™ Adaptaquin Available on GSA/VA Contract for Federal Government customers only.
HIF-prolyl hydroxylase-2 (PHD2) inhibitor; antioxidant
$157.90 - $658.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
59-531-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5953/10
10mg Each for $157.90
Add to cart
59-535-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5953/50
Each for $658.50
Add to cart
Description
Description
HIF-prolyl hydroxylase-2 (PHD2) inhibitor. Blocks glutamate induced ROS production in HT-22 cells, independent of MnSOD. Exhibits neuroprotective effects and enhances functional recovery in rodent intracerebral hemorrhage models, via inhibition of ATF4 dependent genes. Brain penetrant. Also antioxidant.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Safety and Handling
Safety and Handling
Recommended Storage : Store at −20°C