Learn More
Fisherbrand™ Adapter with 1/2 in. NPT Thread x 5/16 in. Barb - Natural Kynar
SDP
Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing.
Catalog No. 01-000-059
Add to cart
Description
Description
  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661
  • USP81
  • USP 8
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable
Warranty and Services
Replacement if not satisfied. 12 Month return Policy.
Specifications
Specifications
Compliance Certificates Available
Threaded Adapter
10 Pack
Natural
Natural Kynar
Threaded Tube Connector for Medical, Pharmaceutical, Industrial applications.
Out of direct sunlight, >20°F <100°F