Learn More
Cole-Parmer Cole-Parmer 24 VDC Loop Powered Adapter For Preamplied pH/orp Cables  DFS Item
Manufacturer:  Cole-Parmer 2700764
Amplify pH or ORP signals without having to replace batteries required for other amplifiers using this extension cable. Recommended for industrial pH, ORP cable runs 30 to 500 feet. It has VAC or VDC powered options.
Catalog No. 50-633-663