Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Adapter for 250 mL Rectangular Bucket in Rotor FA - 6 x 250
Adapts a wide range of centrifuge tubes for use in Rotor FA-6x250.
Supplier: Eppendorf™ 5920768000
Description
- Adapts a wide range of centrifuge tubes to fit in Rotor FA - 6 x 250
- Set of 2 pieces
Specifications
1 x 85 mL Oak Ridge Round Centrifuge Tube | |
For 250 mL rectangular bucket in Rotor FA-6x250 | |
1 | |
Set of 2 pieces |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction