Learn More
Fisherbrand™ Adapter with 3/8 in. NPT Thread x 5/16 in. Barb - Polypropylene
Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing.
Description
- Single & Multi Use Applications
- PVC-Free
- Superior Single Barb Design
- USP Class VI Compliant
- USP 661
- USP81
- USP 8
- RoHS Compliant
- Gamma Stable
- Animal Derivative Free
- Lot Traceable
Warranty and Services
Replacement if not satisfied. 12 Month return Policy.
Specifications
Compliance Certificates Available | |
Non-Animal Derived Polypropylene | |
10 Pack | |
Natural |
Threaded Adapter | |
Threaded Tube Connector for Medical, Pharmaceutical, Industrial applications. | |
Out of direct sunlight, >20°F <100°F |