Learn More
Cole-Parmer Cole-Parmer Adapter, Metrohm meter to BNC electrode  DFS Item
Manufacturer:  Cole-Parmer 0598370
BNC is the most common electrode connector, but some pH meters and electrodes come with different types of connectors. Order an adapter to ensure your meter and electrode are compatible.
Catalog No. 50-633-591