Learn More
Cole-Parmer Parker Hannifin Compression threaded adapter, PFA, 1/4" tubing OD x 1/8" female NPT  DFS Item
Manufacturer:  Cole-Parmer 3132052
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.
Catalog No. 50-631-910