Learn More
Cole-Parmer Parker Hannifin Compression threaded adapter, PFA, 3/8" tubing OD x 1/4" female NPT  DFS Item
Manufacturer:  Cole-Parmer 3132054
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.
Catalog No. 50-631-914