Learn More
Chemglass Life Sciences Style A Thermocouple Transition Adapter, Type "T" (Blue) Male Standard to Female Mini
SDP
Supplier:  Chemglass Life Sciences CG3499A01
Encompass_Preferred
Style A Thermocouple Transition Adapter, Type "T" (Blue) Male Standard to Female Mini. Adapters provide quick and easy connections between standard (OST) and miniature (SMP) connector systems.
Catalog No. CG3499A01
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.