Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541835
Description
Peptide sequence: RLVPCGAAIS WSAVPEQPLD VTANGFPQGT TKKTIGSLQA RSQISKVELF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
This gene is a member of the ADAR (adenosine deaminase acting on RNA) family. Using site-specific adenosine modification, proteins encoded by these genes participate in the pre-mRNA editing of nuclear transcripts. The protein encoded by this gene, tRNA-specific adenosine deaminase 1, is responsible for the deamination of adenosine 37 to inosine in eukaryotic tRNA.
Specifications
ADAT1 | |
Polyclonal | |
Unconjugated | |
ADAT1 | |
Adat1; Adenosine deaminase; adenosine deaminase acting on tRNA; adenosine deaminase tRNA specific 1; adenosine deaminase, tRNA specific 1; adenosine deaminase, tRNA-specific 1; HADAT1; mADAT1; MMADAT1; tRNA-specific 1; tRNA-specific adenosine deaminase 1; tRNA-specific adenosine-37 deaminase; zgc:162299 | |
Rabbit | |
Protein A | |
RUO | |
23536 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BUB4 | |
ADAT1 | |
Synthetic peptide directed towards the C-terminal of human ADAT1 (aa 390-439). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction