Learn More
Selleck Chemical LLCSupplier Diversity Partner Adavivint (SM04690) DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S87615MG
SM04690 is a potent and specific inhibitor of canonical Wnt signaling with an EC50 of 19.5 nM for inhibiting the TCF/LEF reporter. It is ∼150- to 500-fold more potent than the other known Wnt inhibitors across multiple cellular assays.
Catalog No. 50-194-8241