Learn More
Aviva Systems Biology Corp AZIN2 Antibody - middle region (ARP52476_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52476_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ADC. Alternate names: AZI2, KIAA1945, ODC-p, ODC1LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62888