Learn More
Abnova™ ADCK2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00090956T02
Catalog No. 89-932-638
Add to cart
Description
Description
Specifications
Specifications
293T
NM_052853.3
pCMV-ADCK2 full-length
100 μL
Human
ADCK2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 69kDa
Transient overexpression cell lysate was tested with anti-ADCK2 antibody (Mfr. No. H00090956-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot