Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCK4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550307

Catalog No. PA550307


Add to cart

Description

Description

The antibody detects endogenous levels of total ADCK4 protein.

This gene encodes a protein with two copies of a domain found in protein kinases. The encoded protein has a complete protein kinase catalytic domain, and a truncated domain that contains only the active and binding sites of the protein kinase domain, however, it is not known whether the protein has any kinase activity. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

ADCK4
Polyclonal
Unconjugated
COQ8B
Liquid
COQ8B
Fusion protein corresponding to a region derived from internal residues of human aarF domain containing kinase 4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q96D53
FLJ12229,aarF domain containing kinase 4
Rabbit
IgG
100 μL
-20°C
Primary
79934
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit