Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenine, >98%, Affymetrix/USB™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
73-24-5
C5H5N5
135.13
MFCD00041790
GFFGJBXGBJISGV-UHFFFAOYSA-N
adenine,1h-purin-6-amine,6-aminopurine,9h-purin-6-amine,vitamin b4,adenin,adeninimine,leuco-4,6-amino-1h-purine,6-amino-3h-purine
190
CHEBI:16708
7H-purin-6-amine
C1=NC2=C(N1)C(=NC=N2)N
73-24-5
C5H5N5
135.13
MFCD00041790
GFFGJBXGBJISGV-UHFFFAOYSA-N
adenine,1h-purin-6-amine,6-aminopurine,9h-purin-6-amine,vitamin b4,adenin,adeninimine,leuco-4,6-amino-1h-purine,6-amino-3h-purine
190
CHEBI:16708
7H-purin-6-amine
C1=NC2=C(N1)C(=NC=N2)N
Melting Point >360°C (decomposition)
Packaging Plastic bottle
Quantity 1kg
UN Number UN2811
Formula Weight 135.13
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material Adenine