Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adeno-associated virus 8 Mouse anti-Adenovirus, Unconjugated, Clone: HL2381, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $380.00

Specifications

Antigen Adeno-associated virus 8
Applications Dot Blot, ELISA, Neutralization
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF23051MI
View Documents
MilliporeSigma
MABF2305100UL
100 μL
Each for $380.00
Add to cart
 
MABF23052MI
View Documents
MilliporeSigma
MABF230525UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

Adeno-associated virus 8 Monoclonal antibody specifically detects Adeno-associated virus 8 in Adenovirus samples. It is validated for Dot Blot, ELISA, Neutralizing.
Specifications

Specifications

Adeno-associated virus 8
Unconjugated
Mouse
Adeno-associated virus serotype 8;AAV8
Monoclonal
Protein G Purified
Dot Blot, ELISA, Neutralization
Purified
Adenovirus
Recombinant Adeno-associated virus serotype 8 capsid protein.
Primary
Stable for 1 year at -20°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit