Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adeno-associated Virus 9 Mouse anti-Virus, Unconjugated, Clone: HL2374, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen Adeno-associated Virus 9
Applications Dot Blot, ELISA, Neutralization
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF23261MI
View Documents
MilliporeSigma
MABF2326100UG
100 μg
Each for $372.00
Add to cart
 
MABF23262MI
View Documents
MilliporeSigma
MABF232625UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

Adeno-associated Virus 9 Monoclonal antibody specifically detects Adeno-associated Virus 9 in Virus samples. It is validated for Dot Blot, ELISA, Neutralizing.
Specifications

Specifications

Adeno-associated Virus 9
Unconjugated
Mouse
AAV9;Adeno-associated viruses serotype 9
Monoclonal
Protein G Purified
Dot Blot, ELISA, Neutralization
Purified
Virus
Adeno-associated virus 9 capsids.
Primary
Stable for 1 year at -20°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit