Learn More
Cell Biolabs IncSupplier Diversity Partner ADENO TITER STAINING KT TRL SZ  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK109T
The QuickTiter Adenovirus Titer Immunoassay measures functional titer in less time with greater accuracy than traditional plaque-forming unit assays. Results in 2.5 days vs. 10 days with PFU assays. 20 assays/kit.
Catalog No. 50-673-752