Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Adenosine 5'-diphosphate, disodium salt hydrate, 98%

Catalog No. p-3756535
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

CAS: 16178-48-6 | C10H13N5Na2O10P2 | 471.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16178-48-6
Molecular Formula C10H13N5Na2O10P2
Molecular Weight (g/mol) 471.17
MDL Number MFCD00150927
InChI Key ORKSTPSQHZNDSC-WCYUCLFNNA-L
Synonym adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate
PubChem CID 85315
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Absorbance 0.76 to 0.80 (A250/A260), 0.15 to 0.17 (A280/A260)
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
Solubility Information (5% w/v in water) Clear colorless
Spectrophotometry 97.5 to 102.5% (ADP-Na2H,on dry basis) (calculated) 82% min. (ADP,on as is basis) (lambda max. 260nm) molar absorptivity 15000+/-2%,pH 7
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
Formula Weight 471.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Adenosine 5′-diphosphate, disodium salt hydrate
Show More Show Less
EINECSNumber 240-314-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.