Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine 5'-monophosphate sodium salt hydrate, 99%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
149022-20-8
C10H13N5NaO7P·xH2O
369.21
FYWLYWMEGHCZAX-MCDZGGTQSA-M
sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate x:1:x,adenosine 5-monophosphoric acid disodium salt,adenosine 5'-monophosphate sodium salt,sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate,pubchem14181,sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate 2:1:x,5'-adenylic acid,sodium salt, hydrate 9ci,disodium hydrate adenosine-5-monophosphate 2-
23674992
sodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate
C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)[O-])O)O.[Na+]
149022-20-8
C10H13N5NaO7P·xH2O
369.21
FYWLYWMEGHCZAX-MCDZGGTQSA-M
sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate x:1:x,adenosine 5-monophosphoric acid disodium salt,adenosine 5'-monophosphate sodium salt,sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate,pubchem14181,sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate 2:1:x,5'-adenylic acid,sodium salt, hydrate 9ci,disodium hydrate adenosine-5-monophosphate 2-
23674992
sodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate
C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)[O-])O)O.[Na+]
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 369.21
Physical Form Powder
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Water 20% Max.
Color White
pH 4.50 to 6.50
Quantity 1g
Chemical Name or Material Adenosine 5′-monophosphate sodium salt hydrate
Show More Show Less