Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5′-triphosphate Disodium Salt, 99%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
987-65-5
987-65-5
Packaging Amber Glass Bottle
Quantity 5 g
CAS Max % 1
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.