Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine 5'-triphosphate disodium salt hydrate Thermo Scientific™

Click to view available options
Quantity:
5g
10g
25g
100g
500g
Packaging:
Plastic bottle
34369-07-8
C10H12N5O13P3
503.15
MFCD00150754
ZKHQWZAMYRWXGA-KQYNXXCUSA-J
adenosine 5'-triphosphate disodium salt trihydrate,adenosine 5'-triphosphate disodium salt,adenosine 5'-triphosphate disodium salt trihydrate, disodium salt
134129625
({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(phosphonatooxy)phosphinate
NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
34369-07-8
C10H12N5O13P3
503.15
MFCD00150754
ZKHQWZAMYRWXGA-KQYNXXCUSA-J
adenosine 5'-triphosphate disodium salt trihydrate,adenosine 5'-triphosphate disodium salt,adenosine 5'-triphosphate disodium salt trihydrate, disodium salt
134129625
({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(phosphonatooxy)phosphinate
NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Formula Weight 605.19
Physical Form Crystalline Powder
Packaging Plastic bottle
Quantity 500g
Chemical Name or Material Adenosine-5'-Triphosphate Disodium Salt Trihydrate