Learn More
Adenosine A2aR Mouse anti-Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit, Sheep, FITC, Clone: 7F6-G5-A2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP139474F
Catalog No. NBP139474F
Add to cart
Description
Description
Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
Specifications
Adenosine A2aR
7F6-G5-A2
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
ADORA2A
Full length recombinant human Adenosine A2a R. [UniProt# P29274]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
FITC
PBS with 0.05% Sodium Azide
adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8
Mouse
IgG2a
0.1 ml
Apoptosis, GPCR, Immunology, Innate Immunity
Primary
135