Learn More
Adenosine A2aR Mouse anti-Human, Alexa Fluor 405, Clone: 599717, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB94971V100UG
Catalog No. FAB9497V100
Add to cart
This item is not returnable. View return policy
Description
Description
Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Adenosine A2aR
599717
Flow Cytometry 0.25-1 ug/10^6 cells
P29274
A2aR, adenosine A2 receptor, Adenosine A2a R, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, ADORA2A, hA2aR, RDC8
NS0 mouse myeloma cell line transfected with human Adenosine A2a R Met1-Ser412 Accession # P29274
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Adenosine A2a R in HEK293 human cell line transfected with Human Adenosine A2a R and not irrelavant transfectants in flow cytometry.
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Supernatant
Mouse
IgG2a
100 ug
Primary
135
Human